DCPIP assay reagent
Leverandør: Cayman Chemical
700942-700
CAYM700942-700EA 805 NOK
CAYM700942-700
DCPIP assay reagent
Assay ELISAs
The vial contains 700 μg of 2,6-dichlorophenolindophenol (DCPIP) assay reagent dissolved in UltraPure water. Thaw the reagent prior to use in the MitoCheck® Complex II Activity Assay Kit.