Calcium assay kit
Leverandør: Cayman Chemical
701220-192
CAYM701220-192EA 7375 NOK
CAYM701220-192
Calcium assay kit
Assay Cellulær analyse
For the determination of total calcium concentration in a variety of biological samples, as well as tissue homogenates and cell lysates.
  • Measure calcium in plasma, serum, urine, tissue homogenates, and cell lysates
  • Assay 40 samples in duplicate
  • Measure calcium down to 0,25 mg/dl
  • Plate-based colorimetric measurement (570 to 590 nm)
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Påvisningsgrense 0.25 mg/dl