Astaxanthine ≥97%, crystalline solid

Leverandør: Cayman Chemical

Synonymer: Carophyll Pink, (3S,3'S)-Astaxanthin, Natupink, (6S,6'S)-3,3'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(6-hydroxy-2,4,4-trimethylcyclohex-2-enone), BioAstin

70685-5 70685-25
CAYM70685-5EA 1900 NOK
CAYM70685-5 CAYM70685-25
Astaxanthine ≥97%, crystalline solid
Astaxanthine

Astaxanthini is a carotenoid pigment found primarily in marine animals including shrimp and salmon. It is a potent lipid-soluble antioxidant.

Formel: C₄₀H₅₂O₄
MW: 596.852 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00672621
CAS-nummer: 472-61-7

Bestill nå

Spesifikasjon Testresultater

Purity ≥97%
λmax 473 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR