S-Methylisothiouronium sulphate

Leverandør: Cayman Chemical

Synonymer: S-Methylisothiourea sulphate, 2-Methyl-2-thiopseudourea sulphate, S-Methylthiouronium sulphate, 2-Methyl-2-thiopseudourea hemisulphate

81300-1
CAYM81300-1EA 830 NOK
CAYM81300-1
S-Methylisothiouronium sulphate
S-Methylisothiouronium sulphate
Formel: C₄H₁₂N₄S₂·H₂SO₄
MDL nummer: MFCD00013055
CAS-nummer: 867-44-7
EINECS: 212-759-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR