Hydrogen and High Purity Air Generator for GC-FID
Leverandør: LUCIDITY
928276
CCDL928276EA 0 NOK
CCDL928276
Hydrogen and High Purity Air Generator for GC-FID
Gassgeneratorer Hydrogengeneratorer
Generator that produces all necessary gasses required to run the Lucidity GC-FID.
  • Hydrogen and high purity air generator for the Lucidity GC-FID
  • Small footprint and economical
  • Self-installable instrument that only requries DI-H₂O
The Lucidity GAS produces hydrogen and high purity air. The economical GAS with its small footprint compliments the Lucidity GC-FID perfectly. The GAS produces all necessary gasses required to run the Lucidity GC-FID. Just add DI-H2O to the self-installable instrument and your lab can have access to gasses required forMore Product Information