Wound cleansers and plasters, Salvequick
Leverandør: Cederroth / Orkla Care AB
1880
CEDE1880EA 0 NOK
CEDE1880
Wound cleansers and plasters, Salvequick
Førstehjelpsutstyr Bandasjer
Wound cleansers and adhesive bandages (plasters) packed in a PE plastic zip bag. The wound cleansers are intended for mechanical cleansing of minor wounds. The adhesive bandages are intended to be used as mechanical barrier and for absorption of exudates from minor wounds.
  • Wound cleansers: Sterile tissues soaked in 0,9% NaCl
  • Adhesive bandages: Micro perforated PE foil coated with polyacrylate adhesive; non woven wound pad (viscose and PP/PE) with a non stick wound contact layer (PE); wrapped in cold seal paper
Adhesive bandages: 25×72 mm