Elastic bandage with clips
Leverandør: Cederroth / Orkla Care AB
1882
CEDE1882EA 1300 NOK
CEDE1882
Elastic bandage with clips
Førstehjelpsutstyr Bandasjer
This elastic bandage with fastening clip is intended to be used for strong fixation bandaging, for support and relief bandaging, for fixation of a wound dressing, and for aftercare of distortion, dislocations, contusions, and fractures. Made of 65% cotton and 35% polyamide.
Not intended for use directly on open wounds.