Burn gel dressing/face mask
Leverandør: Cederroth / Orkla Care AB
51011014
CEDE51011014EA 0 NOK
CEDE51011014
Burn gel dressing/face mask
Førstehjelpsutstyr Spray mot forbrenning
Large burn gel dressing provides rapid cooling and effective pain relief for first and second degree burns.
  • Sterile dressing prevents injury from penetrating deeper into skin
  • Cold reduces pain and swelling
  • Can also be used as a face mask
  • Easy to use with clear instructions
Suitable for burns on larger areas such as legs, arms, or chest. For use as a face mask, simply tear off the parts of the dressing in the pre-cut holes for eyes, nose and mouth.