Sterile burn gel dressing
Leverandør: Cederroth / Orkla Care AB
51011015
CEDE51011015EA 1350 NOK
CEDE51011015
Sterile burn gel dressing
Førstehjelpsutstyr Spray mot forbrenning
Burn gel dressing provides rapid cooling and effective pain relief for first and second degree burns.
  • Sterile dressing prevents injury from penetrating deeper into skin
  • Cold reduces pain and swelling
  • Shelf life of 5 years
Easy to use with clear instructions. Suitable for use on burns up to 20x20 cm. For larger areas use several dressings.