Canister filling stand
Leverandør: CEMO
8007 10100
CEMG8007EA 1850 NOK
CEMG8007 CEMG10100
Canister filling stand
Stativer
Filling stands, made from highly resistant stainless steel, suitable for acids and lyes.
  • Facilitates emptying of canisters up to 60 L
  • Cage secures mounting of canisters
  • Precise metering by swivelling of the canister