Drum filling station
Leverandør: CEMO
8674
CEMG8674EA 7000 NOK
CEMG8674
Drum filling station
Tønnelagring
This filling station is suitable as a storage and filling station for two 200 L barrels and with additional drum pallet for four 200 L drums.
  • Made from high-quality polyethylene
  • High chemical resistance
  • Can be driven under
  • Collection volume: 450 L