Face screen without carrier, S591
Leverandør: CENTURION SAFETY PRODUCTS
S591
CENUS591EA 370 NOK
CENUS591
Face screen without carrier, S591
Ansiktsskjermer
Long acetate face screen, to be combined with the carrier S54 for helmet attachment.
  • Resistant against organic and inorganic chemicals
  • Ideal for metal cutting applications
Length: 210 mm
Sertifiseringer: EN 166