Welding face shield
Leverandør: CENTURION SAFETY PRODUCTS
S903NCE
CENUS903NCEEA 1300 NOK
CENUS903NCE
Welding face shield
Ansiktsskjermer
This lightweight helmet-mounted welding shield is highly impact resistant.
  • Shield is made of robust Polyamide PA6 (Nylon)
  • Supplied with Polycarbonate chipping lens
  • Compatible with all Centurion helmets except for the Spartan helmet
Window: 110×90 mm
Sertifiseringer: EN 175