Visual comparator test kit for dssolved oxygen by the indiigo carmine method, CHEMets
Leverandør: CHEMetrics
K-7512
CHETK-7512EA 2375 NOK
CHETK-7512
Visual comparator test kit for dssolved oxygen by the indiigo carmine method, CHEMets
Miljø testkit
Test kits for environmental and drinking water applications in the ppm range.
  • Visual indication of dissolved oxygen by formation of blue product
  • Method is not to interferences from temperature, salinity, or dissolved gases such as sulfide
  • Results are expressed as ppm (mg/L) O₂
Leveringsinformasjon: Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, comparator, 25 ml sample cup and instructions