Toluidin blått

Leverandør: CHROMA

Synonymer: Toluidine blue O zinc chloride double salt

1B-481
CHRO1B-481EA 0 NOK
CHRO1B-481
Toluidin blått
Toluidin blått
Formel: C₁₅H₁₆ClN₃S·0.5ZnCl₂
MW: 373.97 g/mol
MDL nummer: MFCD03548293
CAS-nummer: 6586-04-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR