Azure B (azure I)

Leverandør: CHROMA

Synonymer: C.I. 52010, N,N,N′-Trimethylthionin, Azure B, Trimethylthionine chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride, Methylene Azur B

1B-487
CHRO1B-487EA 0 NOK
CHRO1B-487
Azure B (azure I)
Azure B (azure I)
Formel: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011935
CAS-nummer: 531-55-5
EINECS: 208-511-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR