Anti-6X His tag Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: AD1.1.10]

Leverandør: Columbia Biosciences
AP-1711
COBSAP-1711EA 7375 NOK
COBSAP-1711
Anti-6X His tag Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: AD1.1.10]
Antistoffer
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol EGLN2
Antigen-navn egl-9 family hypoxia-inducible factor 2
Environmentally Preferable
Antigen synonymer HIFPH1|HIF-PH1|EIT6|HPH-1|HPH-3|PHD1
Antistofftype Tag
Klonalitet Monoclonal
Konjugering AP (Alkaline Phosphatase)
Klon AD1.1.10
Reaktivitet
Vert Mouse
Gen-ID 112398
Isotype IgG1 kappa
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR