Anti-c-Myc tag Mouse Monoclonal Antibody (DyLight® 650) [clone: 9E10]

Leverandør: Columbia Biosciences
D10-1714-100
COBSD10-1714-100EA 7375 NOK
COBSD10-1714-100
Anti-c-Myc tag Mouse Monoclonal Antibody (DyLight® 650) [clone: 9E10]
Antistoffer
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR