Anti-HA tag Rabbit Polyclonal Antibody (DyLight® 650)

Leverandør: Columbia Biosciences
D10-1830
COBSD10-1830EA 7375 NOK
COBSD10-1830
Anti-HA tag Rabbit Polyclonal Antibody (DyLight® 650)
Antistoffer
Type: Tag
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol HA
Environmentally Preferable
Antigen-navn HA tag
Antistofftype Tag
Klonalitet Polyclonal
Konjugering DyLight® 650
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR