Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: Columbia Biosciences
D3-103
COBSD3-103EA 4525 NOK
COBSD3-103
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR