Anti-IgA Goat Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: Columbia Biosciences
D3-110-A
COBSD3-110-AEA 4525 NOK
COBSD3-110-A
Anti-IgA Goat Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR