Anti-IgG1 Goat Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: Columbia Biosciences
D3-112-1
COBSD3-112-1EA 4525 NOK
COBSD3-112-1
Anti-IgG1 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR