Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: Columbia Biosciences
D3-112-M
COBSD3-112-MEA 2350 NOK
COBSD3-112-M
Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Type: Secondary
Antigen: IgM (Mouse) (mouse IgM)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol IgM
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering APC (Allophycocyanin)
Reaktivitet Mouse
Vert Goat
Isotype IgG
ELISA Yes
Flowcytometri Yes
Kyssabsorpsjon Bovine, Rabbit
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR