Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: M2]

Leverandør: Columbia Biosciences
D3-1718-100 D3-1718-1MG
COBSD3-1718-100EA 7375 NOK
COBSD3-1718-100 COBSD3-1718-1MG
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: M2]
Antistoffer
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR