Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: M2]

Leverandør: Columbia Biosciences
D3-1718-100 D3-1718-1MG
COBSD3-1718-100EA 7375 NOK
COBSD3-1718-100 COBSD3-1718-1MG
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: M2]
Antistoffer
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol DYKDDDDK
Environmentally Preferable
Antigen-navn DYKDDDDK tag
Antistofftype Tag
Klonalitet Monoclonal
Konjugering APC (Allophycocyanin)
Klon M2
Reaktivitet
Vert Mouse
Isotype IgG1 kappa
ELISA Yes
Flowcytometri Yes
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR