Anti-EP4 receptor C-terminal amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI) Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: Columbia Biosciences
D3-1866-50
COBSD3-1866-50EA 6775 NOK
COBSD3-1866-50
Anti-EP4 receptor C-terminal amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI) Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Type: Primary
Antigen: EP4 receptor C-terminal amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn EP4 receptor
Antistofftype Primary
Klonalitet Polyclonal
Konjugering APC (Allophycocyanin)
Reaktivitet
Vert Rabbit
Isotype IgG
ELISA Yes
Flowcytometri Yes
Epitop C-terminal
Aminosyretall 459 to 488
Sekvens GSGRAGPAPKGSSLQVTFPSETLNLSEKCI
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR