Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Leverandør: Columbia Biosciences
D5-103
COBSD5-103EA 4525 NOK
COBSD5-103
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antistoffer
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering RPE (R-Phycoerythrin)
Reaktivitet Chicken
Vert Goat
Isotype IgG
ELISA Yes
Flowcytometri Yes
Kyssabsorpsjon Bovine, Mouse, Rabbit
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR