Anti-IgE Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Leverandør: Columbia Biosciences
D5-110-E
COBSD5-110-EEA 7375 NOK
COBSD5-110-E
Anti-IgE Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antistoffer
Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR