Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Leverandør: Columbia Biosciences
D5-112-FAB
COBSD5-112-FABEA 4525 NOK
COBSD5-112-FAB
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antistoffer
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR