Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Leverandør: Columbia Biosciences
D5-114-1MG D5-114-100
COBSD5-114-1MGEA 7300 NOK
COBSD5-114-1MG COBSD5-114-100
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antistoffer
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR