Mouse IgG2a Isotype Control (RPE (R-Phycoerythrin))

Leverandør: Columbia Biosciences
D5-1602A
COBSD5-1602AEA 4525 NOK
COBSD5-1602A
Mouse IgG2a Isotype Control (RPE (R-Phycoerythrin))
Antistoffer
Type: Control
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: None
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antistofftype Control
Klonalitet Polyclonal
Konjugering RPE (R-Phycoerythrin)
Reaktivitet
Vert Mouse
Isotype IgG2a
ELISA Yes
Flowcytometri Yes
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR