Anti-S Tag Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Leverandør: Columbia Biosciences
D5-1831
COBSD5-1831EA 7375 NOK
COBSD5-1831
Anti-S Tag Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Antistoffer
Type: Tag
Antigen: S Tag (S tag)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol S
Environmentally Preferable
Antigen-navn S tag
Antistofftype Tag
Klonalitet Polyclonal
Konjugering RPE (R-Phycoerythrin)
Reaktivitet
Vert Rabbit
Isotype IgG
ELISA Yes
Flowcytometri Yes
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR