Anti-CB1 receptor Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Leverandør: Columbia Biosciences
D5-1861
COBSD5-1861EA 7375 NOK
COBSD5-1861
Anti-CB1 receptor Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Antistoffer
Type: Primary
Antigen: CB1 receptor
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol CB1
Antigen-navn CB1 receptor
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering RPE (R-Phycoerythrin)
Reaktivitet
Vert Rabbit
Isotype IgG
ELISA Yes
Flowcytometri Yes
Epitop C-terminal
Aminosyretall 461 to 472
Sekvens MSVSTDTSAEAL
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR