Anti-PGIS Rabbit Polyclonal Antibody RPE (R-Phycoerythrin)

Leverandør: Columbia Biosciences
D5-1867
COBSD5-1867EA 7375 NOK
COBSD5-1867
Anti-PGIS Rabbit Polyclonal Antibody RPE (R-Phycoerythrin)
Antistoffer
Type: Primary
Antigen: PGIS sequence amino acids 475-490 (PEFDLSRYGFGLMQPE)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR