Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (DyLight® 550) [clone: M2]

Leverandør: Columbia Biosciences
D9-1718
COBSD9-1718EA 7375 NOK
COBSD9-1718
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (DyLight® 550) [clone: M2]
Antistoffer
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR