Anti-S Tag Rabbit Polyclonal Antibody (DyLight® 550)

Leverandør: Columbia Biosciences
D9-1831
COBSD9-1831EA 7375 NOK
COBSD9-1831
Anti-S Tag Rabbit Polyclonal Antibody (DyLight® 550)
Antistoffer
Type: Tag
Antigen: S Tag (S tag)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR