Anti-T7 Tag Rabbit Polyclonal Antibody (DyLight® 550)

Leverandør: Columbia Biosciences
D9-1832
COBSD9-1832EA 7375 NOK
COBSD9-1832
Anti-T7 Tag Rabbit Polyclonal Antibody (DyLight® 550)
Antistoffer
Type: Tag
Antigen: T7 Tag (T7 tag)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol T7
Environmentally Preferable
Antigen-navn T7 tag
Antistofftype Tag
Klonalitet Polyclonal
Konjugering DyLight® 550
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR