Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Columbia Biosciences
HRP-110
COBSHRP-110EA 3450 NOK
COBSHRP-110
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering HRP (Horseradish Peroxidase)
Reaktivitet Human
Vert Goat
Isotype IgG
Western blot Yes
ELISA Yes
Immunutfelling Yes
Kyssabsorpsjon Bovine, Mouse, Rabbit
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR