Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Columbia Biosciences
HRP-110-FC
COBSHRP-110-FCEA 3450 NOK
COBSHRP-110-FC
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR