Anti-HA tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 16B12]

Leverandør: Columbia Biosciences
HRP-1722
COBSHRP-1722EA 7375 NOK
COBSHRP-1722
Anti-HA tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 16B12]
Antistoffer
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: 16B12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR