Single Use Oral Syringes, 3-Parts
Leverandør: CODAN
62.4960 62.4980 62.6960
CODA62.4960EA 0 NOK
CODA62.4960 CODA62.4980 CODA62.6960
Single Use Oral Syringes, 3-Parts
Sprøyter Sprøyter, medisinsk
The non-sterile oral syringe program consist of reliable, precise, high-quality single use syringes, includes a range with oral tip dispensers with clear and amber barrel for UV light protection of sensitive medications.
  • Three-part syringe with thin silicone ring
  • Easy and very even withdrawal and emptying
  • Smooth plunger guidance
  • Reliable plunger stop
  • Precise scale with dual scale printing
  • Environmentally friendly and PVC-free materials
  • Latex-free
  • Balanced portfolio with centric and eccentric connections
The oral syringes also meets the highestMore Product Information