Cleanroom papers, CRB18
Leverandør: CONNECTICUT CLEANROOM
PB2C001WHITE
CONNPB2C001WHITEEA 6325 NOK
CONNPB2C001WHITE
Cleanroom papers, CRB18
Papir
CRB-18 cleanroom papers are saturated with syntheic polymers which seals individual fibers into the sheet to minimize particle generation. The papers can be used for a variety of uses in the cleanroom, primarily note taking and record keeping.
  • Heat resistant for use in laser printers and photocopy machines
  • Autoclavable