Twin Bucket
Leverandør: CONTEC
2743S
CONT2743SEA 8050 NOK
CONT2743S
Twin Bucket
Bøtter
Twin Bucket is an autoclavable 20 L divided bucket made of durable PP with a curved stainless steel wringer.
  • Ideal for smaller controlled environments and labs with limited storage space
  • Designed to work with the curved stainless steel wringer
  • Four castors for added mobility
  • Fully autoclavable (except castors)
One side of the bucket can be used for fresh cleaning and disinfecting solutions, and the other one for waste.
Leveringsinformasjon: Supplied with lid, stainless steel wringer and four castors.