Cleanroom wipes, Polywipe Light
Leverandør: CONTEC
7070-6025
CONT7070-6025EA 9425 NOK
CONT7070-6025
Cleanroom wipes, Polywipe Light
Papirservietter Renromsservietter Tørkepapir, renrom
Dry wipes designed for durability and ease of use that are ideal for cage and equipment wipe down or downtime 'super cleaning'.
  • Polyester interlock knit
  • Knife-cut edge
  • Product is double bagged to ensure product integrity
  • Lot numbers provide traceability
  • Economical bulk packaging
The Polywipe Light wipe is an unlaundered standardweight polyester interlock knit wipe with knife-cut edges that is to be used in less than critical applications (in environments of ISO Class 6 and higher). The wipes have the same basic functional characteristics as launderedMore Product Information