Cleanroom wipes, Amplitude™ VIP-L
Leverandør: CONTEC
AMVL0001 AMVL0002 AMVL0004
CONTAMVL0001EA 12650 NOK
CONTAMVL0001 CONTAMVL0002 CONTAMVL0004
Cleanroom wipes, Amplitude™ VIP-L
Papirservietter Renromsservietter Tørkepapir, renrom
This standard weight wipe with textured surface is knitted from filament polyester. It is ideal for effective cleaning.
  • Soft textured wipe cleans surfaces without scratching
  • Extremely low in both particles and fibers
  • Good sorbency with solvents
  • Chemical resistant
The wipe is laser-cut, which seals the fibers at the edges. It is laundered and packaged in an ISO Class 4 cleanroom to further reduce contamination.