Bond tack cloth
Leverandør: CONTEC
BS20CB-12
CONTBS20CB-12EA 8575 NOK
CONTBS20CB-12
Bond tack cloth
Papirservietter Rengjøringskluter for gjenbruk Kluter for følsomme oppgaver
These polyester cloths are ideal for the surface preparation prior to painting. Also suitable for particle removal in printed circuit board manufacturing and for eliminating solvents in some applications.
  • Custom blended tack formula for optimal adhesive texture, transfer resistance, heat resistance and anti-static performance
  • Sealed edge technology minimises particles and fibres
  • "Napped" surface easily removes dirt particles
  • Water-based tack is applied at high heat under pressure