Cleanroom sorbent pillows and socks, EliminatR®
Leverandør: CONTEC
ELRPIL
CONTELRPILEA 5225 NOK
CONTELRPIL
Cleanroom sorbent pillows and socks, EliminatR®
Materialer for håndtering av søl
Disposable cleanroom sorbent pillows and socks are designed for large spill pickup or as a drip-catching pad under equipment. They consist of 100% polyester knitted fabric with an durable two-ply polyester outer covering. The pillow is designed for large spill pickup and leak control. The acid-resistant sock is ideal for placement around machinery or drums.
  • Excellent sorbency for aqueous spills
  • Superior holding ability for oils and acids
Pillows and socks are free of any cellulosic contaminants, and are recommended for use in ISO Class 6 or higherMore Product Information