Mopping systems, EasyCurve™
Leverandør: CONTEC
2724 EZSE0100 EZSE0150 EZMP0350 EZMP0300
CONT2724EA 2900 NOK
CONT2724 CONTEZSE0100 CONTEZSE0150 CONTEZMP0350 CONTEZMP0300
Mopping systems, EasyCurve™
Mopper
Contec’s EasyCurve™ Mop was designed specifically for critical and controlled environments, now available with a ribbed microfiber face giving users smooth and easy mopping for better particle pick up and contamination removal. The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability, making the EasyCurve™ an ideal cleaning tool for ceilings, walls and floors. Available in nonsterile and validated sterile versions.
Mop heads:
Very absorbent and drip-free
Compatible with most common solutions, solvents andMore Product Information