Disinfectant spray, Contec® HydroPure
Leverandør: CONTEC
FBC56HP FBT16HP
CONTFBC56HPEA 3825 NOK
CONTFBC56HP CONTFBT16HP
Disinfectant spray, Contec® HydroPure
Rengjøringsløsninger Desinfeksjon
Contec® HydroPure a blend of 6% stabilised hydrogen peroxide and purified water and leaves little to no residue and is ideal for use on most surfaces. Contec® HydroPure is BPR authorised at EU level and is efficacious against bacteria, fungi, moulds and yeasts.
  • Hydrogen peroxide breaks down to water and oxygen making it very low residue
  • Filtered to 0,2 μm under Grade A airflow in a Grade B cleanroom ensuring the product is free from contamination and particulates
  • Tigger spray and 'bag in bottle' protected system prevents bottle contents from becoming