Cleanroom wipes, sterile, Polynit Heatseal
Leverandør: CONTEC
PN-99IR LWPS0007 LWPS0006 LWPS1004
115-2145EA 18925 NOK
115-2145 115-1862 CONTLWPS0006 CONTLWPS1004
Cleanroom wipes, sterile, Polynit Heatseal
Papirservietter Renromsservietter Desinfiseringsservietter, renrom
100% no-run interlock knit polyester wipes with sealed edges.
  • Validated sterile to a 10⁻⁶ SAL
  • Laundered knitted 100% polyester fabric for extremely low levels of particles and fibres
  • Extremely low levels of particles, fibres and residue
  • Laser-cut, sealed edge prevents release of fibres
  • Laundered and packaged in an ISO Class 4 cleanroom
Sterile Polynit Heatseal wipes are made of 100% knitted polyester that is laser cut to bond the fibres at the edges of the wipe. This creates a very clean non abrasive edge that allows full utilisation of the wipeMore Product Information
Bestill nå
SPESIFIKASJONER
Modell Polynit Polynit Heatseal Polynit Heatseal
Environmentally Preferable
grunnvekt 140 g/m² 140 g/m² 140 g/m²
Absorberingsevne 384 ml/m²
Absorberingsevne - utvendig 330 ml/m²
Absorberingsevne - innvendig 2,74 ml/g 2,43 ml/g
Absorberingsevne - hastighet 1 s <1 s
Ekstraherbare stoffer - bruk av deionisert vann 0,009 g/m² 0,008 g/m² 0,008 g/m²
Ekstraherbare stoffer - bruk av isopropylalkohol 0,005 g/m² 0,007 g/m² 0,007 g/m²
Fibre (>100 µm) 1,54×10³/m² 0,142×10³/m² 0,142×10³/m²
Partikkelgenerering(>=0.5 µm) 1,53 × 10⁶/m² 2,60 × 10⁶/m² 2,60 × 10⁶/m²
Ioner - klorid 0,10 0,003 ppm 0,003 ppm
Ioner - natrium 0,13 0,180 ppm 0,180 ppm